Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCG5
Entrez Gene ID
508224
Gene Name
secretogranin V
Gene Aliases
7B2, Sgne1
Chromosome Location
Chr.10: 30003358 - 30062438 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045998.2 NP_001039463.1 3 - 4 412 87
GenBank mRNA BC105453.1 - 3 - 4 413 87

View Full Product Details