Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REM1
Entrez Gene ID
508364
Gene Name
RRAD and GEM like GTPase 1
Gene Aliases
-
Chromosome Location
Chr.13: 61074023 - 61083230 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046001.2 NP_001039466.1 1 - 2 58 97
GenBank mRNA BC114085.1 - 1 - 2 59 97

View Full Product Details