Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRAF2
Entrez Gene ID
508703
Gene Name
PRA1 domain family member 2
Gene Aliases
-
Chromosome Location
Chr.X: 87153478 - 87156113 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046009.2 NP_001039474.1 1 - 2 188 76
GenBank mRNA BC112850.1 - 1 - 2 190 76

View Full Product Details