Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RND1
Entrez Gene ID
508869
Gene Name
Rho family GTPase 1
Gene Aliases
-
Chromosome Location
Chr.5: 30923453 - 30930386 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046016.1 NP_001039481.1 4 - 5 529 85
GenBank mRNA BC113283.1 - 4 - 5 529 85

View Full Product Details