Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D31
Entrez Gene ID
511062
Gene Name
TBC1 domain family member 31
Gene Aliases
WDR67
Chromosome Location
Chr.14: 16631180 - 16698179 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046073.2 NP_001039538.1 6 - 7 917 140
GenBank mRNA BC114127.1 - 6 - 7 918 140

View Full Product Details