Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFAF3
Entrez Gene ID
511968
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 3
Gene Aliases
C22H3orf60, C3orf60
Chromosome Location
Chr.22: 50968487 - 50972548 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046105.2 NP_001039570.1 4 - 5 459 95
GenBank mRNA BC105350.1 - 4 - 5 460 95

View Full Product Details