Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SH3GL1
Entrez Gene ID
512719
Gene Name
SH3 domain containing GRB2 like 1, endophilin A2
Gene Aliases
-
Chromosome Location
Chr.7: 19706807 - 19734112 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046130.2 NP_001039595.1 5 - 6 662 89
GenBank mRNA BC105444.1 - 5 - 6 663 89
BT025352.1 - 5 - 6 545 89

View Full Product Details