Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATG101
Entrez Gene ID
513049
Gene Name
autophagy related 101
Gene Aliases
C12orf44, C5H12orf44
Chromosome Location
Chr.5: 27808218 - 27814091 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046134.1 NP_001039599.1 2 - 3 759 149
GenBank mRNA BC105530.1 - 2 - 3 759 149

View Full Product Details