Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLBD2
Entrez Gene ID
514347
Gene Name
phospholipase B domain containing 2
Gene Aliases
P76
Chromosome Location
Chr.17: 61041378 - 61063716 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046170.1 NP_001039635.1 7 - 8 1165 68
GenBank mRNA BC112461.1 - 7 - 8 1165 68

View Full Product Details