Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP1LC3A
Entrez Gene ID
514547
Gene Name
microtubule associated protein 1 light chain 3 alpha
Gene Aliases
-
Chromosome Location
Chr.13: 63918608 - 63920647 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046175.1 NP_001039640.1 1 - 2 184 72
GenBank mRNA BC113348.1 - 1 - 2 184 72

View Full Product Details