Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNHIT1
Entrez Gene ID
514997
Gene Name
zinc finger HIT-type containing 1
Gene Aliases
-
Chromosome Location
Chr.25: 35473360 - 35479721 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046185.2 NP_001039650.1 1 - 2 53 111
GenBank mRNA BC114191.1 - 1 - 2 54 111

View Full Product Details