Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAPB
Entrez Gene ID
517728
Gene Name
NSF attachment protein beta
Gene Aliases
-
Chromosome Location
Chr.13: 42038671 - 42076724 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046233.2 NP_001039698.1 6 - 7 560 85
GenBank mRNA BC114061.1 - 6 - 7 563 85

View Full Product Details