Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A1BG
Entrez Gene ID
518955
Gene Name
alpha-1-B glycoprotein
Gene Aliases
-
Chromosome Location
Chr.18: 65641101 - 65645263 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046243.2 NP_001039708.1 2 - 3 127 109
GenBank mRNA BC105374.1 - 2 - 3 130 109

View Full Product Details