Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL44
Entrez Gene ID
532389
Gene Name
mitochondrial ribosomal protein L44
Gene Aliases
-
Chromosome Location
Chr.2: 112068965 - 112076913 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046321.2 NP_001039786.1 1 - 2 196 117
GenBank mRNA BC112531.1 - 1 - 2 198 117

View Full Product Details