Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM34
Entrez Gene ID
539820
Gene Name
tripartite motif containing 34
Gene Aliases
-
Chromosome Location
Chr.15: 47890399 - 47904766 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046461.2 NP_001039926.1 6 - 7 1042 138
GenBank mRNA BC114124.1 - 6 - 7 1047 138

View Full Product Details