Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NSRP1
Entrez Gene ID
539841
Gene Name
nuclear speckle splicing regulatory protein 1
Gene Aliases
CCDC55, NSrp70
Chromosome Location
Chr.19: 21237745 - 21283421 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046462.1 NP_001039927.1 5 - 6 540 85
GenBank mRNA BC112694.1 - 5 - 6 540 85

View Full Product Details