Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NANS
Entrez Gene ID
540602
Gene Name
N-acetylneuraminate synthase
Gene Aliases
-
Chromosome Location
Chr.8: 63192956 - 63212667 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046482.1 NP_001039947.1 5 - 6 951 55
GenBank mRNA BC114646.1 - 5 - 6 951 55

View Full Product Details