Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADCYAP1
Entrez Gene ID
615187
Gene Name
adenylate cyclase activating polypeptide 1
Gene Aliases
PACAP
Chromosome Location
Chr.24: 35732908 - 35739931 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046555.1 NP_001040020.1 3 - 4 343 133
GenBank mRNA AY924308.1 - 3 - 4 343 133
BC123523.1 - 4 - 5 133

View Full Product Details