Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPA1
Entrez Gene ID
280701
Gene Name
inorganic pyrophosphatase 1
Gene Aliases
PP
Chromosome Location
Chr.28: 26418192 - 26456501 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075118.1 NP_001068586.1 1 - 2 139 111
GenBank mRNA BC114891.1 - 1 - 2 139 111
M95283.1 - 1 - 2 125 111

View Full Product Details