Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2B6
Entrez Gene ID
504769
Gene Name
cytochrome P450 subfamily 2B
Gene Aliases
CYP2B
Chromosome Location
Chr.18: 50290689 - 50308034 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075173.1 NP_001068641.1 4 - 5 510 127
GenBank mRNA BC112659.1 - 4 - 5 510 127

View Full Product Details