Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR3GL
Entrez Gene ID
505641
Gene Name
RNA polymerase III subunit G like
Gene Aliases
-
Chromosome Location
Chr.3: 21396710 - 21415301 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075213.1 NP_001068681.1 2 - 3 246 92
GenBank mRNA BC114765.1 - 2 - 3 246 92

View Full Product Details