The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001075279.1 | NP_001068747.1 | 2 - 3 | 181 | 67 |
GenBank mRNA | BC118272.1 | - | 2 - 3 | 181 | 67 |