Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPP1
Entrez Gene ID
510998
Gene Name
membrane palmitoylated protein 1
Gene Aliases
-
Chromosome Location
Chr.X: 36176490 - 36209890 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075484.1 NP_001068952.1 11 - 12 1359 73
GenBank mRNA BC118256.1 - 11 - 12 1359 73

View Full Product Details