Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYSMD2
Entrez Gene ID
511013
Gene Name
LysM domain containing 2
Gene Aliases
-
Chromosome Location
Chr.10: 58609344 - 58642587 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075485.1 NP_001068953.1 1 - 2 283 133
GenBank mRNA BC116100.1 - 1 - 2 283 133

View Full Product Details