Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHPT1
Entrez Gene ID
511291
Gene Name
choline phosphotransferase 1
Gene Aliases
-
Chromosome Location
Chr.5: 65511727 - 65543536 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075507.1 NP_001068975.1 2 - 3 558 111
GenBank mRNA BC116046.1 - 2 - 3 558 111

View Full Product Details