Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHPT1
Entrez Gene ID
511291
Gene Name
choline phosphotransferase 1
Gene Aliases
-
Chromosome Location
Chr.5: 65511727 - 65543536 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075507.1 NP_001068975.1 7 - 8 1198 130
GenBank mRNA BC116046.1 - 7 - 8 1198 130

View Full Product Details