Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRG1
Entrez Gene ID
512533
Gene Name
FSHD region gene 1
Gene Aliases
-
Chromosome Location
Chr.27: 19178454 - 19192323 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075566.1 NP_001069034.1 7 - 8 761 124
GenBank mRNA BC114781.1 - 7 - 8 761 124

View Full Product Details