Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO2
Entrez Gene ID
512589
Gene Name
F-box protein 2
Gene Aliases
-
Chromosome Location
Chr.16: 42028287 - 42034351 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075569.1 NP_001069037.1 3 - 4 926 84
GenBank mRNA BC118302.1 - 3 - 4 926 84

View Full Product Details