Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO2
Entrez Gene ID
512589
Gene Name
F-box protein 2
Gene Aliases
-
Chromosome Location
Chr.16: 42028287 - 42034351 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075569.1 NP_001069037.1 4 - 5 1019 56
GenBank mRNA BC118302.1 - 4 - 5 1019 56

View Full Product Details