Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAF1
Entrez Gene ID
514182
Gene Name
nuclear assembly factor 1 ribonucleoprotein
Gene Aliases
-
Chromosome Location
Chr.6: 2620826 - 2671246 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075652.2 NP_001069120.2 1 - 2 401 103
GenBank mRNA BC114778.1 - 1 - 2 402 103
DV822960.1 - 1 - 2 328 103
DV845783.1 - 1 - 2 402 103

View Full Product Details