Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAF1
Entrez Gene ID
514182
Gene Name
nuclear assembly factor 1 ribonucleoprotein
Gene Aliases
-
Chromosome Location
Chr.6: 2620826 - 2671246 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075652.2 NP_001069120.2 6 - 7 965 153
GenBank mRNA BC114778.1 - 6 - 7 966 153
CK949061.1 - 3 - 4 277 153

View Full Product Details