Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ISCU
Entrez Gene ID
514789
Gene Name
iron-sulfur cluster assembly enzyme
Gene Aliases
-
Chromosome Location
Chr.17: 64357116 - 64363222 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075683.2 NP_001069151.1 3 - 4 405 72
GenBank mRNA BC118409.1 - 3 - 4 406 72

Gene Symbol
LOC533308
Entrez Gene ID
533308
Gene Name
iron-sulfur cluster scaffold homolog (E. coli)-like
Gene Aliases
-
Chromosome Location
Chr.10: 51529607 - 51530234 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243316.1 NP_001230245.1 1 - 1 72
GenBank mRNA BC149555.1 - 1 - 1 72

View Full Product Details