Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPCD
Entrez Gene ID
516908
Gene Name
deleted in primary ciliary dyskinesia homolog
Gene Aliases
-
Chromosome Location
Chr.26: 22334762 - 22356876 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075745.2 NP_001069213.1 3 - 4 303 70
GenBank mRNA BC114010.1 - 3 - 4 304 70

View Full Product Details