Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTERF3
Entrez Gene ID
526138
Gene Name
mitochondrial transcription termination factor 3
Gene Aliases
MTERFD1
Chromosome Location
Chr.14: 67961055 - 67981699 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075881.1 NP_001069349.1 3 - 4 904 115
GenBank mRNA BC116050.1 - 3 - 4 904 115

View Full Product Details