Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BEGAIN
Entrez Gene ID
527886
Gene Name
brain enriched guanylate kinase associated
Gene Aliases
-
Chromosome Location
Chr.21: 65422663 - 65468748 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075902.2 NP_001069370.2 1 - 2 37 68
GenBank mRNA BC114844.1 - 1 - 2 38 68

View Full Product Details