The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001075982.1 | NP_001069450.1 | 3 - 4 | 142 | 88 |
GenBank mRNA | BC118239.1 | - | 3 - 4 | 142 | 88 |