Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMTOR3
Entrez Gene ID
533155
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
Gene Aliases
MAP2K1IP1, MAPKSP1
Chromosome Location
Chr.6: 24787040 - 24798441 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075982.1 NP_001069450.1 4 - 5 201 131
GenBank mRNA BC118239.1 - 4 - 5 201 131

View Full Product Details