Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COQ10A
Entrez Gene ID
538821
Gene Name
coenzyme Q10A
Gene Aliases
-
Chromosome Location
Chr.5: 57065731 - 57069113 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076130.2 NP_001069598.1 2 - 3 443 81
GenBank mRNA BC114817.1 - 2 - 3 444 81

View Full Product Details