Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSRNP3
Entrez Gene ID
540832
Gene Name
cysteine and serine rich nuclear protein 3
Gene Aliases
FAM130A2
Chromosome Location
Chr.2: 30608229 - 30812957 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076244.1 NP_001069712.1 3 - 4 468 98
GenBank mRNA BC116047.1 - 3 - 4 468 98

View Full Product Details