Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COLEC11
Entrez Gene ID
613991
Gene Name
collectin subfamily member 11
Gene Aliases
-
Chromosome Location
Chr.8: 110960692 - 110983690 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076303.2 NP_001069771.1 2 - 3 246 116
GenBank mRNA BC118358.1 - 2 - 3 247 116

View Full Product Details