Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM216B
Entrez Gene ID
614187
Gene Name
family with sequence similarity 216 member B
Gene Aliases
C12H13orf30, C13orf30
Chromosome Location
Chr.12: 12923398 - 12933177 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076315.1 NP_001069783.1 3 - 4 639 63
GenBank mRNA BC118252.1 - 3 - 4 639 63

View Full Product Details