Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTARC2
Entrez Gene ID
615506
Gene Name
mitochondrial amidoxime reducing component 2
Gene Aliases
MARC2, MOSC2
Chromosome Location
Chr.16: 24224873 - 24273784 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076380.2 NP_001069848.1 2 - 3 505 68
GenBank mRNA BC116001.1 - 2 - 3 506 68

View Full Product Details