Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
HSPB11
Entrez Gene ID
616194
Gene Name
heat shock protein family B (small) member 11
Gene Aliases
C3H1ORF41
Chromosome Location
Chr.3: 92262412 - 92337726 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076412.1 NP_001069880.1 4 - 5 372 99
GenBank mRNA BC118487.1 - 4 - 5 372 99

View Full Product Details