The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001076412.1 | NP_001069880.1 | 6 - 7 | 483 | 138 |
GenBank mRNA | BC118487.1 | - | 6 - 7 | 483 | 138 |