Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OGFR
Entrez Gene ID
767947
Gene Name
opioid growth factor receptor
Gene Aliases
-
Chromosome Location
Chr.13: 54566992 - 54574602 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077019.1 NP_001070487.1 4 - 5 389 79
GenBank mRNA BC105559.1 - 4 - 5 389 79

View Full Product Details