Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
B3GALT4
Entrez Gene ID
768038
Gene Name
beta-1,3-galactosyltransferase 4
Gene Aliases
-
Chromosome Location
Chr.23: 7451341 - 7453548 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077097.1 NP_001070565.1 1 - 2 30 156
GenBank mRNA BC113234.1 - 1 - 2 30 156

View Full Product Details