Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACSF2
Entrez Gene ID
768237
Gene Name
acyl-CoA synthetase family member 2
Gene Aliases
-
Chromosome Location
Chr.19: 36241654 - 36273156 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078112.1 NP_001071580.1 12 - 13 1547 61
GenBank mRNA BC118331.1 - 12 - 13 1547 61

View Full Product Details