Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FGF12
Entrez Gene ID
615823
Gene Name
fibroblast growth factor 12
Gene Aliases
-
Chromosome Location
Chr.1: 74675841 - 75288328 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079789.1 NP_001073257.1 3 - 4 297 61
GenBank mRNA BC118169.1 - 3 - 4 297 61

View Full Product Details