Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFAP4
Entrez Gene ID
286766
Gene Name
microfibril associated protein 4
Gene Aliases
-
Chromosome Location
Chr.19: 34082571 - 34085108 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080217.1 NP_001073686.1 2 - 3 109 122
GenBank mRNA BC113263.1 - 2 - 3 109 122

View Full Product Details