Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLAMF1
Entrez Gene ID
281489
Gene Name
signaling lymphocytic activation molecule family member 1
Gene Aliases
SLAM
Chromosome Location
Chr.3: 8969591 - 9008637 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174184.4 NP_776609.2 2 - 3 501 129
GenBank mRNA AF329970.1 - 2 - 3 440 129
BC114833.1 - 2 - 3 502 129

View Full Product Details